missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nitrite Nitrogen Standard, 100 ppm N (328.5 ppm NO2-), Ricca Chemical
Specifications
Specifications
| Packaging | Amber Glass Bottle |
| CAS Min % | 98.0, 0.06 |
| CAS Max % | 100.0, 0.06 |
| Physical Form | Liquid |
| Color | Colorless |
| pH | Approximately 7 |
| Boiling Point | 100°C |
| Quantity | 120 mL |
| Grade | ACS |
| Concentration or Composition (by Analyte or Components) | 1mL = 0.1mg N, 100ppm N as Nitrite (328.5ppm NO2-) |
| Show More |
Safety and Handling
| ShelfLife | 6 months |