missing translation for 'onlineSavingsMsg'
Learn More

Potassium Nitrite, Crystal, ACS, 96%, Spectrum™ Chemical

Catalog No. 18612808
Change view
Click to view available options
Quantity:
25 g
125 g
500 g
2.5 kg
25 kg
Packaging:
Poly Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18612808 25 kg Poly Bottle
18612804 25 g Poly Bottle
18612805 125 g Poly Bottle
18612806 500 g Poly Bottle
18612807 2.5 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18612808 Supplier Spectrum Chemical Mfg Cor Supplier No. P135325KGBL
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7758-09-0

Specifications

CAS Max % 1
Physical Form Crystals or Needles
Quantity 25 kg
Molecular Weight (g/mol) 85.10
Percent Purity 96%
Grade ACS/Reagent
Packaging Poly Bottle
Chemical Name or Material Potassium Nitrite

CAUTION: For manufacturing, processing or repacking. Read and understand the label and Safety Data Sheet (SDS) prior to use.