missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nitrite, MilliporeSigma™ Supelco™
Specifications
Specifications
| Chemical Name or Material | Nitrite |
| CAS | 7758-09-0 |
| Type | Certified Reference Material |
| Quantity | 2 mL |
| Grade | Certified Reference Material |