missing translation for 'onlineSavingsMsg'
Learn More

Potassium nitrite, 96+%, ACS reagent

Catalog No. AC423065000
Change view
Click to view available options
Quantity:
5 g
500 g
2.5 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC423065000 500 g
AC423060050 5 g
AC423060025 2.5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC423065000 Supplier Thermo Scientific Chemicals Supplier No. 423065000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7758-09-0 | KNO2 | 85.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7758-09-0
Molecular Formula KNO2
Molecular Weight (g/mol) 85.10
MDL Number MFCD00011408
InChI Key BXNHTSHTPBPRFX-UHFFFAOYSA-M
Synonym potassium nitrite, nitrous acid, potassium salt, caswell no. 698, ccris 3959, potassium nitrite 1:1, epa pesticide chemical code 076203, nitrous acid, potassium salt 1:1, kaliumnitrit, acmc-1bi01, dsstox_cid_22320
PubChem CID 516910
SMILES [K+].[O-]N=O

Specifications

Melting Point 387.0°C
Color White to Yellow
Physical Form Crystals
Quantity 500 g
Packaging Plastic bottle
Assay Percent Range 96+%
Linear Formula KNO2
Fieser 10,329
Merck Index 15, 7770
Solubility Information Solubility in water: 3000g/L water (20°C). Other solubilities: 413 g/100mL water (100°C), slightly soluble in alcohol, soluble in hot alcohol, very soluble in liquid nh3
IUPAC Name potassium;nitrite
Formula Weight 85.1
Percent Purity ≥96%
Grade ACS Reagent
Chemical Name or Material Potassium nitrite, 96+%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Toxic if swallowed.
Very toxic to aquatic life.
May intensify fire; oxidizer.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 231-832-4
RTECSNumber TT3750000
TSCA TSCA

RUO – Research Use Only