missing translation for 'onlineSavingsMsg'
Learn More

Nitrite - WP, Concentrate for dilution to 1 L, MilliporeSigma™ Supelco™

Catalog No. 111002595
Change view
Click to view available options
Quantity:
2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111002595 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111002595 Supplier Merck Emd Millipore Supplier No. PE11532ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This proficiency testing sample is produced in accordance with ISO/IEC 17043:2010. All information regarding the use of this material can be found in the reporting packet supplied for each sample.

Specifications

Packaging Ampule
CAS 7758-09-0
Type Proficiency Testing Material
Quantity 2 mL