CAS RN 63-74-1
Sulfanilamide Certified ACS, Fisher Chemical
CAS: 63-74-1 Molecular Formula: C6H8N2O2S Molecular Weight (g/mol): 172.202 MDL Number: MFCD00007939 InChI Key: FDDDEECHVMSUSB-UHFFFAOYSA-N Synonym: 4-Aminobenzenesulfonamide,P-Anilinesulfonamide PubChem CID: 5333 ChEBI: CHEBI:45373 IUPAC Name: 4-aminobenzenesulfonamide SMILES: C1=CC(=CC=C1N)S(=O)(=O)N
Thermo Scientific Chemicals Sulfanilamide 98%
CAS: 63-74-1 Molecular Formula: C6H8N2O2S Molecular Weight (g/mol): 172.202 MDL Number: MFCD00007939 InChI Key: FDDDEECHVMSUSB-UHFFFAOYSA-N Synonym: 4-Aminobenzenesulfonamide PubChem CID: 5333 ChEBI: CHEBI:45373 IUPAC Name: 4-aminobenzenesulfonamide SMILES: C1=CC(=CC=C1N)S(=O)(=O)N
Sulfanilamide, 98%
CAS: 63-74-1 Molecular Formula: C6H8N2O2S Molecular Weight (g/mol): 172.202 MDL Number: MFCD00007939 InChI Key: FDDDEECHVMSUSB-UHFFFAOYSA-N Synonym: sulfanilamide,sulphanilamide,sulfamine,sulphonamide,p-aminobenzenesulfonamide,sulfonylamide,sulfonamide,p-aminobenzenesulfamide,bacteramid,streptasol PubChem CID: 5333 ChEBI: CHEBI:45373 IUPAC Name: 4-aminobenzenesulfonamide SMILES: C1=CC(=CC=C1N)S(=O)(=O)N
Sulfanilamide Melting Point Standard, Pharmaceutical Secondary Standard, Certified Reference Material, MilliporeSigma™ Supelco™
Sulfanilamide Melting Point Standard also called Sulfanilamide is an antibacterial drug.
Sulfanilamide Reagent, 1% (w/v) in 10% (v/v) Hydrochloric Acid, Ricca Chemical
CAS: 63-74-1 Molecular Formula: C6H8N2O2S Molecular Weight (g/mol): 172.202 InChI Key: FDDDEECHVMSUSB-UHFFFAOYSA-N Synonym: sulfanilamide,sulphanilamide,sulfamine,sulphonamide,p-aminobenzenesulfonamide,sulfonylamide,sulfonamide,p-aminobenzenesulfamide,bacteramid,streptasol PubChem CID: 5333 ChEBI: CHEBI:45373 IUPAC Name: 4-aminobenzenesulfonamide SMILES: C1=CC(=CC=C1N)S(=O)(=O)N
Sulfanilamide, For Nitrogen (Nitrate/Nitrite), 1% (w/v), LabChem™
CAS: 63-74-1 Molecular Formula: C6H8N2O2S Molecular Weight (g/mol): 172.20 InChI Key: FDDDEECHVMSUSB-UHFFFAOYSA-N Synonym: sulfanilamide,sulphanilamide,sulfamine,sulphonamide,p-aminobenzenesulfonamide,sulfonylamide,sulfonamide,p-aminobenzenesulfamide,bacteramid,streptasol PubChem CID: 5333 ChEBI: CHEBI:45373 IUPAC Name: 4-aminobenzene-1-sulfonamide SMILES: NC1=CC=C(C=C1)S(N)(=O)=O
Sulfanilamide 99.0+%, TCI America™
CAS: 63-74-1 Molecular Formula: C6H8N2O2S Molecular Weight (g/mol): 172.202 MDL Number: MFCD00007939 InChI Key: FDDDEECHVMSUSB-UHFFFAOYSA-N Synonym: sulfanilamide,sulphanilamide,sulfamine,sulphonamide,p-aminobenzenesulfonamide,sulfonylamide,sulfonamide,p-aminobenzenesulfamide,bacteramid,streptasol PubChem CID: 5333 ChEBI: CHEBI:45373 IUPAC Name: 4-aminobenzenesulfonamide SMILES: C1=CC(=CC=C1N)S(=O)(=O)N
Sulfanilamide, 100.26%, MP Biomedicals™
CAS: 63-74-1 Molecular Formula: C6H8N2O2S Molecular Weight (g/mol): 172.202 InChI Key: FDDDEECHVMSUSB-UHFFFAOYSA-N Synonym: sulfanilamide,sulphanilamide,sulfamine,sulphonamide,p-aminobenzenesulfonamide,sulfonylamide,sulfonamide,p-aminobenzenesulfamide,bacteramid,streptasol PubChem CID: 5333 ChEBI: CHEBI:45373 IUPAC Name: 4-aminobenzenesulfonamide SMILES: C1=CC(=CC=C1N)S(=O)(=O)N
Sulfanilamide 99.0+%, TCI America™
CAS: 63-74-1 Molecular Formula: C6H8N2O2S Molecular Weight (g/mol): 172.202 MDL Number: MFCD00007939 InChI Key: FDDDEECHVMSUSB-UHFFFAOYSA-N Synonym: sulfanilamide,sulphanilamide,sulfamine,sulphonamide,p-aminobenzenesulfonamide,sulfonylamide,sulfonamide,p-aminobenzenesulfamide,bacteramid,streptasol PubChem CID: 5333 ChEBI: CHEBI:45373 IUPAC Name: 4-aminobenzenesulfonamide SMILES: C1=CC(=CC=C1N)S(=O)(=O)N
Sulfanilamide 99.9+%, TCI America™
CAS: 63-74-1 Molecular Formula: C6H8N2O2S Molecular Weight (g/mol): 172.202 MDL Number: MFCD00007939 InChI Key: FDDDEECHVMSUSB-UHFFFAOYSA-N Synonym: sulfanilamide,sulphanilamide,sulfamine,sulphonamide,p-aminobenzenesulfonamide,sulfonylamide,sulfonamide,p-aminobenzenesulfamide,bacteramid,streptasol PubChem CID: 5333 ChEBI: CHEBI:45373 IUPAC Name: 4-aminobenzenesulfonamide SMILES: C1=CC(=CC=C1N)S(=O)(=O)N
Sulfanilamide, Reagent Grade, >98.0%, LabChem™
CAS: 63-74-1 Molecular Formula: C6H8N2O2S Molecular Weight (g/mol): 172.202 InChI Key: FDDDEECHVMSUSB-UHFFFAOYSA-N Synonym: sulfanilamide,sulphanilamide,sulfamine,sulphonamide,p-aminobenzenesulfonamide,sulfonylamide,sulfonamide,p-aminobenzenesulfamide,bacteramid,streptasol PubChem CID: 5333 ChEBI: CHEBI:45373 IUPAC Name: 4-aminobenzenesulfonamide SMILES: C1=CC(=CC=C1N)S(=O)(=O)N
Sulfanilamide, Reagent Grade, Ricca Chemical
CAS: 63-74-1 Molecular Formula: C6H8N2O2S Molecular Weight (g/mol): 172.202 InChI Key: FDDDEECHVMSUSB-UHFFFAOYSA-N PubChem CID: 5333 ChEBI: CHEBI:45373 IUPAC Name: 4-aminobenzenesulfonamide SMILES: C1=CC(=CC=C1N)S(=O)(=O)N
Sulfanilamide 98+%, Reagents
CAS: 63-74-1 Molecular Formula: C6H8N2O2S Molecular Weight (g/mol): 172.20 InChI Key: FDDDEECHVMSUSB-UHFFFAOYSA-N Synonym: Sulfamine, 4-Aminobenzenesulfonamide IUPAC Name: 4-aminobenzene-1-sulfonamide SMILES: NC1=CC=C(C=C1)S(N)(=O)=O
Sulfanilamide, 99.0- 101.0%, Spectrum™ Chemical
CAS: 63-74-1 Molecular Weight (g/mol): 172.21