missing translation for 'onlineSavingsMsg'
Learn More

Sulfanilamide, For Nitrogen (Nitrate/Nitrite), 1% (w/v), LabChem™

Catalog No. LC254502
Change view
Click to view available options
Quantity:
500 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC254502 1 L
LC254501 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. LC254502 Supplier LabChem Supplier No. LC254502

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 63-74-1 , 7647-01-0 , 7732-18-5
Molecular Formula C6H8N2O2S
Molecular Weight (g/mol) 172.20
InChI Key FDDDEECHVMSUSB-UHFFFAOYSA-N
Synonym sulfanilamide, sulphanilamide, sulfamine, sulphonamide, p-aminobenzenesulfonamide, sulfonylamide, sulfonamide, p-aminobenzenesulfamide, bacteramid, streptasol
PubChem CID 5333
ChEBI CHEBI:45373
IUPAC Name 4-aminobenzene-1-sulfonamide
SMILES NC1=CC=C(C=C1)S(N)(=O)=O

Specifications

Color Colorless
CAS Max % 1, 4.34, 94.66
Linear Formula C6H8N2O2S
UN Number UN1789
Solubility Information Soluble in water
Formula Weight 172.2
Percent Purity 1% (w/v)
Identification Passes Test
Packaging Amber Poly
Decomposition Information Hydrogen chloride; Carbon dioxide; Carbon monoxide; Nitrogen oxides; Sulfur compounds
Quantity 1 L
Physical Form Liquid
Chemical Name or Material Sulfanilamide, For Nitrogen (Nitrate/Nitrite)
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Do not breathe mist, vapors, spray.
Wear protective gloves/eye protection.
Wash exposed skin thoroughly after handling.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Recommended Storage Room Temperature