missing translation for 'onlineSavingsMsg'
Learn More

Sulfanilamide 99.0+%, TCI America™

Catalog No. S08335G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S08335G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S08335G Supplier TCI America Supplier No. S08335G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 63-74-1
Molecular Formula C6H8N2O2S
Molecular Weight (g/mol) 172.202
MDL Number MFCD00007939
InChI Key FDDDEECHVMSUSB-UHFFFAOYSA-N
Synonym sulfanilamide, sulphanilamide, sulfamine, sulphonamide, p-aminobenzenesulfonamide, sulfonylamide, sulfonamide, p-aminobenzenesulfamide, bacteramid, streptasol
PubChem CID 5333
ChEBI CHEBI:45373
IUPAC Name 4-aminobenzenesulfonamide
SMILES C1=CC(=CC=C1N)S(=O)(=O)N

Specifications

Melting Point 166°C
Color White
Quantity 5 g
Formula Weight 172.20
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Sulfanilamide [for Biochemical Research]
EINECSNumber (3)-1973&(3)-2179
RTECSNumber WO8400000
TSCA Yes