missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACS certifié sulfanilamide, Fisher Chemical
Spécifications
Spécifications
| Catégorie | Sulfanilamide |
| Nom ou substance chimique | Sulfanilamide |
| Forme physique | Solide |
| pH | 5.8 à 6.1 |
| Point de fusion | 163 °C |
| Emballage | Bouteille en verre |
| Couleur | Blanc |
| Quantité | 100 g |
| Fourchette de pourcentages d’essai | ≥98% |
| Formule linéaire | 4-NH2C6H4SO2NH2 |
| Afficher plus de résultats |
Sécurité et manipulation
| Hazard Statement | WARNING! |
| Precautionary Statement | Emergency Overview Irritating to eyes, respiratory system and skin. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Obtain medical attention. Do not induce vomiting. Obtain medical attention. . . NFPA Health:2 Flammability:1 Instability:1 |