missing translation for 'onlineSavingsMsg'
Learn More

Sulfanilamide, Reagent Grade, Ricca Chemical

Catalog No. RDCS116025A
Change view
Click to view available options
Package: Natural Poly; Size: 2.5 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
RDCS116025A Package: Natural Poly; Size: 2.5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RDCS116025A Supplier Ricca Chemical Company Supplier No. RDCS11602.5A5

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 63-74-1
CAS Min % 98.0
CAS Max % 100.0
Packaging Natural Poly Bottle
Color White
Quantity 2.5 kg
Molecular Formula C6H8N2O2S
InChI Key FDDDEECHVMSUSB-UHFFFAOYSA-N
SMILES C1=CC(=CC=C1N)S(=O)(=O)N
IUPAC Name 4-aminobenzenesulfonamide
Molecular Weight (g/mol) 172.202
PubChem CID 5333
ChEBI CHEBI:45373
Grade Reagent
Chemical Name or Material Sulfanilamide
Show More Show Less