missing translation for 'onlineSavingsMsg'
Learn More

Sulfanilamide Melting Point Standard, Pharmaceutical Secondary Standard, Certified Reference Material, MilliporeSigma™ Supelco™

Catalog No. 111015008
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015008 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015008 Supplier Merck Emd Millipore Supplier No. PHR10931G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Sulfanilamide Melting Point Standard also called Sulfanilamide is an antibacterial drug.

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Specifications

CAS 63-74-1
Physical Form Neat
Linear Formula H2NC6H4SO2NH2
MDL Number MFCD00007939
Melting Point ∼166°C; 164°C to 166°C
Synonym Sulfanilamide; p-Aminobenzenesulfonamide
Quantity 1 g
Formula Weight 172.20
Grade Certified Reference Material