missing translation for 'onlineSavingsMsg'
Learn More

Sulfanilamide, Reagent Grade, Ricca Chemical

Catalog No. RDCS1160100
Change view
Click to view available options
Quantity:
100 g
Packaging:
White Poly Wide Mouth Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
RDCS1160100 100 g White Poly Wide Mouth Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RDCS1160100 Supplier Ricca Chemical Company Supplier No. RDCS1160100B1

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 63-74-1
Molecular Formula C6H8N2O2S
Molecular Weight (g/mol) 172.202
InChI Key FDDDEECHVMSUSB-UHFFFAOYSA-N
PubChem CID 5333
ChEBI CHEBI:45373
IUPAC Name 4-aminobenzenesulfonamide
SMILES C1=CC(=CC=C1N)S(=O)(=O)N

Specifications

Type Sulfanilamide
Physical Form Powder
CAS Min % 98.0
CAS Max % 100.0
Packaging White Poly Wide Mouth Bottle
Color White
Quantity 100 g
Percent Purity ≥98.0%
Grade Reagent
Chemical Name or Material Sulfanilamide