missing translation for 'onlineSavingsMsg'
Learn More

Sulfanilamide 98+%, Reagents

Catalog No. 01336939
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01336939 100 g
01336938 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01336939 Supplier Labchem Inc Supplier No. C2335200100C2

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Sulfanilamide 98+%, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 63-74-1
Molecular Formula C6H8N2O2S
Molecular Weight (g/mol) 172.20
InChI Key FDDDEECHVMSUSB-UHFFFAOYSA-N
Synonym Sulfamine, 4-Aminobenzenesulfonamide
IUPAC Name 4-aminobenzene-1-sulfonamide
SMILES NC1=CC=C(C=C1)S(N)(=O)=O

Specifications

Melting Point 165.5°C
Color Colorless/White
Melting Range 165.5°C
Packaging Glass Bottle
Quantity 100 g
Specific Gravity 1.08
Grade Reagent
Physical Form Solid
Chemical Name or Material Sulfanilamide 98+%