accessibility menu, dialog, popup

UserName

HNHHHF

CAS RN 12125-01-8

IUPAC Name: amine hydrofluoride
Synonyms: ammonium fluoride
Molecular Weight (g/mol): 37.04
Molecular Formula: FH4N
InChi Key: LDDQLRUQCUTJBB-UHFFFAOYSA-N
SMILES: N.F
Quantity 
  • (5)
  • (4)
  • (2)
  • (2)
  • (4)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (6)
  • (1)
Grade 
  • (6)
  • (3)
  • (3)
  • (3)
  • (2)
  • (3)
  • (7)
  • (5)
Physical Form 
  • (3)
  • (5)
  • (3)
  • (1)
  • (12)
  • (3)
  • (2)
  • (3)
Percent Purity 
  • (12)
  • (4)
  • (1)
  • (1)
  • (3)
  • (7)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (18)
  • (12)
  • (2)
  • (1)
  • (4)
  • (1)
  • (12)
  • (2)
  • (16)
  • (5)
  • (4)
  • (2)
  • (2)
  • (4)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (6)
  • (1)
  • (6)
  • (3)
  • (3)
  • (3)
  • (2)
  • (3)
  • (7)
  • (5)
  • (3)
  • (5)
  • (3)
  • (1)
  • (12)
  • (3)
  • (2)
  • (3)
  • (12)
  • (4)
  • (1)
  • (1)
  • (3)
  • (7)