missing translation for 'onlineSavingsMsg'
Learn More

Sigma Aldrich Ammonium fluoride

Catalog No. 33886925G
Encompass
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
33886925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 33886925G Supplier Sigma Aldrich Supplier No. 33886925G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Ammonium fluoride is employed in a new hydrothermal synthesis of complex flourides NaHoF4 and NaEuF4 these interesting compounds have the fluorite structure and are important for solid-state lasers as well as for photoluminescence and magnetic characteristics.The product was used to prepare Ce3F nanoparticles (NPs) which were further conjugated to photosensitizers along with Au NPs to improve singlet oxygen generation in photosensitizers.

Specifications

CAS 12125-01-8
Quantity 25 g
Molecular Formula H4FN
Linear Formula NH4F
MDL Number MFCD00011423
Molecular Weight (g/mol) 37.04
Percent Purity ≥99.99% trace metals basis