missing translation for 'onlineSavingsMsg'
Learn More

Ammonium fluoride, 96%

Catalog No. AAA1627922
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1627922 100 g
AAA1627936 500 g
AAA162790E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1627922 Supplier Thermo Scientific Chemicals Supplier No. A1627922

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 12125-01-8 | FH4N | 37.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
Ammonium fluoride is used for etching of glass, preservation of food, in printing textile, and as mothproofing agent. It is used in chemical analysis and in brewing. It is used in the synthesis of NaHoF4 and NaEuF4, which are important in solid state lasers as well as photoluminescence. The fluoride ion is used as a base and nucleophile in organic synthesis. Ammonium fluoride providee safe deprotection strategy for the tert-butyldimethylsilyl (TBS) group during RNA synthesis.

Solubility
Freely soluble in water. Slightly soluble in alcohol.

Notes
Hygroscopic and non-cumbustible. On heating, it sublimes and decomposes to hydrogen fluoride and ammonia gas, which can recombine to form ammonium fluoride.

Chemical Identifiers

CAS 12125-01-8
Molecular Formula FH4N
Molecular Weight (g/mol) 37.04
MDL Number MFCD00011423
InChI Key LDDQLRUQCUTJBB-UHFFFAOYSA-N
IUPAC Name amine hydrofluoride
SMILES N.F

Specifications

Quantity 100 g
Linear Formula NH4F
UN Number UN2505
Merck Index 14,522
Solubility Information Freely soluble in water. Slightly soluble in alcohol.
Formula Weight 37.04
Percent Purity 96%
Odor Odorless
Sensitivity Hygroscopic
Density 1.01
Chemical Name or Material Ammonium fluoride
Show More Show Less
Hazard Category H301+H311+H331
Hazard Statement GHS H Statement
H301-H311-H331
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P311-P312-P330-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: AMMONIUM FLUORIDE
EINECSNumber 235-185-9
RTECSNumber BQ6300000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only