missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Fluoride, Crystal, Technical, Spectrum™ Chemical

Catalog No. 18608652
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
45 kg
Packaging:
Poly Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18608652 2.5 kg Poly Bottle
18608648 125 g Poly Bottle
18608651 500 g Poly Bottle
18608649 12 kg Poly Bottle
18608650 45 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18608652 Supplier Spectrum Chemical Mfg Cor Supplier No. A11862.5KG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 12125-01-8

Specifications

CAS Max % 1
Physical Form Crystals and/or Chunks
Quantity 2.5 kg
Molecular Weight (g/mol) 37.04
Grade Technical
Packaging Poly Bottle
Chemical Name or Material Ammonium Fluoride, Crystal

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.