missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium fluoride, Puriss. p.a., ACS Reagent, ≥98%, Solstice
Description
- Leading the Industry For Over 65 Years
- Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
| Physical Form | Solid |
| Quantity | 6 x 1 kg |
| Linear Formula | NH4F |
| UN Number | UN2505 |
| Formula Weight | 37.04 |
| Percent Purity | ≥98% |
| Grade | ACS/puriss. p.a. |
| Residue after Ignition | ≤0.005% (as SO4) |
| Chemical Name or Material | Ammonium Fluoride |
Safety and Handling
| Hazard Statement | Missing Phrase - N15.00950417-P280-P302 + P352 + P312-P304 + P340 + P312-P305 + P351 + P338 + P310 |
| Precautionary Statement | H301 + H311 + H331-H318 |
| DOTInformation | 6.1 / PGIII |
| EINECSNumber | 235-185-9 |