missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Fluoride, MilliporeSigma™
Specifications
Specifications
| Physical Form | Crystals |
| Quantity | 1 kg |
| UN Number | UN2505 |
| Solubility Information | Soluble in water: 820g/L at 20°C |
| Formula Weight | 37.04 |
| Percent Purity | ≥98.0% |
| Grade | ACS |
| Odor | Ammoniacal |
| Packaging | Plastic Bottle |
| Density | 1.01g/cm3 at 20°C |
| Show More |
Safety and Handling
| Hazard Category | H301: Toxic if swallowed |
| Hazard Statement | H311: Toxic in contact with skin |
| Precautionary Statement | 331: Toxic if inhaled |
| DOTInformation | DOT Class: 6.1, Packaging Group: III |
| EINECSNumber | 235-185-9 |
| Recommended Storage | Store at 15° to 25°C (59° to 77°F), Tightly closed. Dry. Keep in well-ventilated place. Keep locked up or in an area accessible only to qualified or authorized persons. |