missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Fluoride, ≥98.0% (F), Solstice

Catalog No. 60047025
Change view
Click to view available options
Quantity:
50 g
Packaging:
Poly bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60047025 50 g Poly bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60047025 Supplier Solstice Supplier No. 5248150G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Eluent additive for LC-MS

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 12125-01-8
Molecular Formula FH4N
Molecular Weight (g/mol) 37.04
MDL Number MFCD00011423
InChI Key LDDQLRUQCUTJBB-UHFFFAOYSA-N
IUPAC Name amine hydrofluoride
SMILES N.F

Specifications

Color Colorless
pH ∼6 (at 20°C)
Physical Form Solid
Quantity 50 g
Linear Formula NH4F
UN Number UN2505
Formula Weight 37.04g/mol
Percent Purity ≥98.0% (F)
Odor Odorless
Packaging Poly bottle
Density ∽ 1.01g/cu.cm at 20°C
Chemical Name or Material Ammonium fluoride, Eluent additive for LC-MS
Show More Show Less
Hazard Statement Missing Phrase - N15.00950417-P280-P302 + P352 + P312-P304 + P340 + P312-P305 + P351 + P338 + P310
DOTInformation 6.1/PGIII
RTECSNumber 235-185-9