missing translation for 'onlineSavingsMsg'
Learn More

Ammonium fluoride, 98+%, ACS reagent

Catalog No. AC423295000
Change view
Click to view available options
Quantity:
5 g
500 g
2.5 kg
Packaging:
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC423295000 500 g Plastic bottle
AC423290050 5 g Plastic bottle
AC423290025 2.5 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC423295000 Supplier Thermo Scientific Chemicals Supplier No. 423295000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 12125-01-8 | FH4N | 37.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 12125-01-8
Molecular Formula FH4N
Molecular Weight (g/mol) 37.04
MDL Number MFCD00011423
InChI Key LDDQLRUQCUTJBB-UHFFFAOYSA-N
IUPAC Name amine hydrofluoride
SMILES N.F

Specifications

Color White
Physical Form Crystalline Powder
Quantity 500 g
Assay Percent Range 98+%
Linear Formula NH4F
Merck Index 15, 519
Solubility Information Solubility in water: soluble. Other solubilities: slightly soluble in alcohol
Formula Weight 37.04
Percent Purity ≥98%
Grade ACS Reagent
Packaging Plastic bottle
Residue after Ignition 0.01% max
Chemical Name or Material Ammonium fluoride
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic in contact with skin.
Toxic if inhaled.
Toxic if swallowed.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air
EINECSNumber 235-185-9
RTECSNumber BQ6300000
TSCA TSCA

RUO – Research Use Only