Learn More
Polysorbate 80
Polysorbate 80, CAS Number-9005-65-6, Polyoxyethylene(20)sorbitan monooleate, 1L, 2mg KOH/g max., Yellow to Amber, 1.0800g/mL, 5ppm max., >110 deg.C, 65 to 80mg KOH/g, Authentic, 15, 7703, MFCD00082107, Glass bottle, Viscous Liquid At 25 deg.C, Indoors +20 deg.C, 1.4710 to 1.4730, 45 to 55mg KOH/g | CAS: 9005-65-6 | (C2H4O)x(C2H4O)z(C2H4O)y(C2H4O)wC24H44O6 | 604.82 g/mol
Supplier: Thermo Scientific Chemicals 278630100
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spécifications
Polysorbate 80 | |
Yellow to Amber | |
1.0800 g/mL | |
(C2H4O)x(C2H4O)z(C2H4O)y(C2H4O)wC24H44O6 | |
Plastic drum | |
CCCCCCCC\C=C/CCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO | |
604.82 | |
Polyoxyethylene(20)sorbitan monooleate |
9005-65-6 | |
Indoors +20°C | |
10 L,10 L | |
MFCD00082107 | |
HDTIFOGXOGLRCB-KTKRTIGZNA-N | |
2-{2-[3,5-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy}ethyl (9E)-octadec-9-enoate | |
Solubility in water: 100g/L. Other solubilities: 100g/L isopropyl alcohol, 10g/L xylene, insoluble in propylene glycol and mineral oil | |
Viscous Liquid |
Identifiants chimiques
9005-65-6 | |
Polyoxyethylene(20)sorbitan monooleate |
RUO – Research Use Only