missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Hypochlorite Solution, 6% available Chlorine, Ricca Chemical
$182.53 - $440.94
Identifiants chimiques
CAS | 7732-18-5 |
---|---|
Molecular Formula | ClNaO |
Molecular Weight (g/mol) | 74.439 |
InChI Key | SUKJFIGYRHOWBL-UHFFFAOYSA-N |
Synonym | sodium hypochlorite, antiformin, sodium oxychloride, chlorox, clorox, javex, javelle water, hypochlorous acid, sodium salt, hypochlorite sodium, carrel-dakin solution |
PubChem CID | 23665760 |
ChEBI | CHEBI:32146 |
IUPAC Name | sodium;hypochlorite |
SMILES | [O-]Cl.[Na+] |
Chemical Identifiers
7732-18-5 | |
74.439 | |
sodium hypochlorite, antiformin, sodium oxychloride, chlorox, clorox, javex, javelle water, hypochlorous acid, sodium salt, hypochlorite sodium, carrel-dakin solution | |
CHEBI:32146 | |
[O-]Cl.[Na+] |
ClNaO | |
SUKJFIGYRHOWBL-UHFFFAOYSA-N | |
23665760 | |
sodium;hypochlorite |
Specifications
Colorless to Yellow | |
92.0,5.88 | |
Liquid | |
ClNaO | |
SUKJFIGYRHOWBL-UHFFFAOYSA-N | |
sodium;hypochlorite | |
23665760 | |
6% | |
6% available Chlorine | |
Sodium Hypochlorite Solution |
7732-18-5,7681-52-9 | |
96.0,6.12 | |
1 L | |
sodium hypochlorite, antiformin, sodium oxychloride, chlorox, clorox, javex, javelle water, hypochlorous acid, sodium salt, hypochlorite sodium, carrel-dakin solution | |
[O-]Cl.[Na+] | |
74.439 | |
CHEBI:32146 | |
Laboratory | |
Amber Poly Bottle |