missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hypochlorite Solution, 6% available Chlorine, Ricca Chemical

Catalog No. 7495732
Change view
Click to view available options
Quantity:
1 L
4 L
Packaging:
Amber Poly Bottle
Black Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
7495732 1 L Amber Poly Bottle
749571 4 L Black Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 7495732 Supplier Ricca Chemical Company Supplier No. 7495.732
Only null left

Chemical Identifiers

CAS 7732-18-5 , 7681-52-9
Molecular Formula ClNaO
Molecular Weight (g/mol) 74.439
InChI Key SUKJFIGYRHOWBL-UHFFFAOYSA-N
Synonym sodium hypochlorite, antiformin, sodium oxychloride, chlorox, clorox, javex, javelle water, hypochlorous acid, sodium salt, hypochlorite sodium, carrel-dakin solution
PubChem CID 23665760
ChEBI CHEBI:32146
IUPAC Name sodium;hypochlorite
SMILES [O-]Cl.[Na+]

Specifications

Color Colorless to Yellow
CAS Min % 92.0, 5.88
CAS Max % 96.0, 6.12
Physical Form Liquid
Quantity 1 L
Percent Purity 6%
Grade Laboratory
Concentration or Composition (by Analyte or Components) 6% available Chlorine
Packaging Amber Poly Bottle
Chemical Name or Material Sodium Hypochlorite Solution