missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Hypochlorite Solution, 6% available Chlorine, Ricca Chemical
Supplier: Ricca Chemical Company 749571
Spécifications
| Sodium Hypochlorite Solution | |
| 7732-18-5,7681-52-9 | |
| 96.0,6.12 | |
| 4 L | |
| sodium hypochlorite, antiformin, sodium oxychloride, chlorox, clorox, javex, javelle water, hypochlorous acid, sodium salt, hypochlorite sodium, carrel-dakin solution | |
| [O-]Cl.[Na+] | |
| 74.439 | |
| CHEBI:32146 | |
| Laboratory | |
| Black Poly Bottle |
| Colorless to Yellow | |
| 92.0,5.88 | |
| Liquid | |
| ClNaO | |
| SUKJFIGYRHOWBL-UHFFFAOYSA-N | |
| sodium;hypochlorite | |
| 23665760 | |
| 6% | |
| 6% available Chlorine |
Identifiants chimiques
| 7732-18-5 | |
| 74.439 | |
| sodium hypochlorite, antiformin, sodium oxychloride, chlorox, clorox, javex, javelle water, hypochlorous acid, sodium salt, hypochlorite sodium, carrel-dakin solution | |
| CHEBI:32146 | |
| [O-]Cl.[Na+] |
| ClNaO | |
| SUKJFIGYRHOWBL-UHFFFAOYSA-N | |
| 23665760 | |
| sodium;hypochlorite |