Learn More
Sartorius Polyethersulfone Membrane Filters
Polyethersulfone (PES) filters have a high internal porosity.
Supplier:  Sartorius 1540747MIN
 Afficher plus de versions de ce produit
Numéro de catalogue. 14-555-457
Il en reste null
Découvrez plus d'offres spéciales
Description
Description
Polyethersulfone (PES) filters have a high internal porosity.
They perform well at high flux with an excellent throughput of aqueous solutions over the entire pH range of 1-14. Thanks to their low non-specific protein adsorption the PES type is recommended for filtering biological and pharmaceutical solutions.
Spécifications
Spécifications
Single-use
Disc
PES
Non-sterile
100/Pk.
Membrane Filter
Membrane Filter
PES
0.2 μm
Recommended for filtering biological and pharmaceutical solutions.
150 μm
47 mm
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS