Learn More
Thermo Scientific™ Pierce™ GST (glutathione-S-transferase)
Pure, lyophilized glutathione S-transferase (GST) protein from Schistosoma japonicum for use in recombinant fusion protein research methods and assays.
Supplier: Thermo Scientific™ 20237
Description
Thermo Scientific™ GST (Glutathione S-Transferase) is pure, lyophilized glutathione S-transferase for use in a variety of protein research procedures.
This pure GST from Schistosoma japonicum is useful in a variety of applications involving GST-tagged fusion proteins and glutathione affinity purification. The expression of foreign proteins in E. coli using a glutathione S-transferase (GST) fusion system is a method of choice for studying the function of particular proteins, for purifying interacting factors and for generating antibodies against fused proteins. Analyses involving GST-tagged proteins usually require or benefit from the use of pure GST as a control or standard.
Highlights:
- Purified from Schistosoma japonicum
- Useful as a molecular weight marker in gel analysis of GST-tagged proteins
- Suitable as a standard or control in ELISA and pull-down assays involving GST-tagged proteins
- Ideal for testing tag-specific cross-reactivity of antibodies in ELISA
- Environmental benefits include:
- Source Reduction
Spécifications
Store at -20°C. | |
Pierce™ | |
Immunoprecipitation | |
GST (glutathione-S-transferase) |
Liquid | |
1 mg | |
Recombinant |
For Research Use Only. Not for use in diagnostic procedures.