missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ TrypLE™ Express Enzyme (1X), no phenol red DFS Item

Unlike porcine trypsin, free of animal-derived components

Supplier:  Gibco™ 12604054

 Afficher plus de versions de ce produit

Numéro de catalogue. 12604054


Il en reste null
Découvrez plus d'offres spéciales

Description

Description

Cleaves peptide bonds on the C-terminal sides of lysine and arginine, and is a direct replacement for trypsin. Exceptional purity increases specificity and reduces damage to cells that can be caused by other enzymes present in some trypsin extracts. Used for dissociating a wide range of adherent mammalian cells, including CHO, HEK 293, A529, primary human keratinocytes, and embryonic stem cells.

  • Gentle on cells
  • Room temperature-stable for 24 months
  • Easy to use
  • Animal origin-free
  • Gentle on cells, purity increases specificity due to the action of a single enzyme and reduces damage caused by cleavage from multiple enzymes in trypsin and other extracts
  • Can be directly substituted for trypsin in existing protocols
  • Dilution alone inactivates TrypLE Express, avoiding the need for trypsin inhibitors, such as FBS

Energy efficient, less hazardous, sustainable packaging

Cell Culture, Mammalian Cell Culture

Greener Choice Claims
  • Environmental benefits include:
    • Source Reduction
    • Énergie renouvelable
Spécifications

Spécifications

EDTA
Mammalian Cells
1 X
Liquid
Sterile-filtered
Energy efficient, Sustainable packaging
Gibco™
Room Temperature Stable
Animal origin-free
Storage conditions: 15°C to 30°C, protect from light
Alternate storage conditions: 2°C to 8°C or -5 to -20°C
Shipping conditions: Room temperature
Shelf life: 24 months from date of manufacture
Animal Origin-free
Adherent Cell Culture
Cell Culture Dissociation Reagent
No Phenol Red
Check and Record
5 L
Room Temperature
In vitro Bioassay
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
FDS
Documents

Documents

Reference Materials

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.