Learn More
Fisherbrand™ ColorFrost™ Plus Microscope Slides
An ideal choice for frozen sections, cytocentrifuge preparations, and standard Papanicolaou smears
 Afficher plus de versions de ce produit
Numéro de catalogue. 22-230-892
Il en reste null
Découvrez plus d'offres spéciales
Description
Description
An ideal choice for frozen sections, cytocentrifuge preparations, and standard Papanicolaou smears
Same outstanding features as regularColorFrost slides with special treatment process that electrostatically adheres tissue sections and cells to the glass without the need for adhesives or protein coatings.
  • Treatment virtually eliminates tissue loss during staining; saves hours of slide preparation
  • Colored marking surface for easy identification throughout procedures
  • Size: 25 × 75mm
Spécifications
Spécifications
Microscope Slide
Yes
25 mm
Glass
75 mm
Burnt Orange
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS
Documents
Documents
Accessoires