Learn More
4-Mercaptophenol, 98%
CAS: 637-89-8 | C6H6OS | 126.17 g/mol
Supplier: Thermo Scientific Chemicals 370630050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spécifications
4-Mercaptophenol | |
637-89-8 | |
100.0 | |
Colorless to Yellow | |
110°C | |
98.5% min. (GC) | |
C6H6OS | |
MFCD00004850 | |
06, IV, 5790 | |
Solubility in water: soluble. Other solubilities: soluble in ethanol,alkali solution,concentrated,sulfuric acid | |
OC1=CC=C(S)C=C1 | |
126.17 | |
126.18 | |
Liquid or Low Melting Solid |
99% | |
98.5 | |
33°C to 35°C | |
149°C to 150°C (25.0 mmHg) | |
Authentic | |
Glass bottle | |
SHC6H4OH | |
5 g | |
4-mercaptophenol, 4-hydroxythiophenol, 4-hydroxybenzenethiol, p-hydroxythiophenol, phenol, 4-mercapto, p-mercaptophenol, thiohydroquinone, monothiohydroquinone, 4-hydroxy thiophenol, hydroquinone, monothio | |
BXAVKNRWVKUTLY-UHFFFAOYSA-N | |
4-sulfanylphenol | |
240147 | |
99% |
Identifiants chimiques
637-89-8 | |
126.17 | |
BXAVKNRWVKUTLY-UHFFFAOYSA-N | |
240147 | |
OC1=CC=C(S)C=C1 |
C6H6OS | |
MFCD00004850 | |
4-mercaptophenol, 4-hydroxythiophenol, 4-hydroxybenzenethiol, p-hydroxythiophenol, phenol, 4-mercapto, p-mercaptophenol, thiohydroquinone, monothiohydroquinone, 4-hydroxy thiophenol, hydroquinone, monothioAfficher plus de résultats | |
4-sulfanylphenol |
Sécurité et manipulation
GHS H Statement
Harmful in contact with skin.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful if inhaled.
GHS P Statement
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 211-307-6
RUO – Research Use Only