missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Mercaptophenol, 98%
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| CAS Min % | 98.5 |
| CAS Max % | 100.0 |
| Melting Point | 33°C to 35°C |
| Color | Colorless to Yellow |
| Boiling Point | 149°C to 150°C (25.0 mmHg) |
| Flash Point | 110°C |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 98.5% min. (GC) |
| Packaging | Glass bottle |
| Linear Formula | SHC6H4OH |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement Harmful in contact with skin. Causes severe skin burns and eye damage. Harmful if swallowed. Harmful if inhaled. |
| Precautionary Statement | GHS P Statement IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear eye protection/face protection. IF IN EYES: Rinse cautiously |
| EINECSNumber | 211-307-6 |
RUO – Research Use Only