CAS RN 5329-14-6
Acid Starch Indicator, Reagents
Acid Starch Indicator, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.
Amidosulfonic acid, 99.99% (metals basis)
CAS: 5329-14-6 Molecular Formula: H3NO3S Molecular Weight (g/mol): 97.088 MDL Number: MFCD00011603 InChI Key: IIACRCGMVDHOTQ-UHFFFAOYSA-N Synonym: amidosulfonic acid,amidosulfuric acid,aminosulfonic acid,imidosulfonic acid,sulfamidic acid,sulfaminic acid,sulphamic acid,jumbo,aminosulfuric acid,sulphamidic acid PubChem CID: 5987 ChEBI: CHEBI:9330 IUPAC Name: sulfamic acid SMILES: NS(=O)(=O)O
Amidosulfonic acid, 98+%
CAS: 5329-14-6 Molecular Formula: H3NO3S Molecular Weight (g/mol): 97.088 MDL Number: MFCD00011603 InChI Key: IIACRCGMVDHOTQ-UHFFFAOYSA-N Synonym: amidosulfonic acid,amidosulfuric acid,aminosulfonic acid,imidosulfonic acid,sulfamidic acid,sulfaminic acid,sulphamic acid,jumbo,aminosulfuric acid,sulphamidic acid PubChem CID: 5987 ChEBI: CHEBI:9330 IUPAC Name: sulfamic acid SMILES: NS(=O)(=O)O
Sulfamic acid, ≥99.5% (alkalimetric), Solstice
CAS: 5329-14-6 Molecular Formula: H3NO3S Molecular Weight (g/mol): 97.088 MDL Number: MFCD00011603 InChI Key: IIACRCGMVDHOTQ-UHFFFAOYSA-N Synonym: amidosulfonic acid,amidosulfuric acid,aminosulfonic acid,imidosulfonic acid,sulfamidic acid,sulfaminic acid,sulphamic acid,jumbo,aminosulfuric acid,sulphamidic acid PubChem CID: 5987 ChEBI: CHEBI:9330 IUPAC Name: sulfamic acid SMILES: NS(=O)(=O)O
Amidosulfonic acid, ACS, 99.3-100.3% (Assay dried basis)
CAS: 5329-14-6 Molecular Formula: H3NO3S Molecular Weight (g/mol): 97.088 MDL Number: MFCD00011603 InChI Key: IIACRCGMVDHOTQ-UHFFFAOYSA-N Synonym: amidosulfonic acid,amidosulfuric acid,aminosulfonic acid,imidosulfonic acid,sulfamidic acid,sulfaminic acid,sulphamic acid,jumbo,aminosulfuric acid,sulphamidic acid PubChem CID: 5987 ChEBI: CHEBI:9330 IUPAC Name: sulfamic acid SMILES: NS(=O)(=O)O
Sulfamic acid, 99%
CAS: 5329-14-6 Molecular Formula: H3NO3S Molecular Weight (g/mol): 97.09 InChI Key: IIACRCGMVDHOTQ-UHFFFAOYSA-N Synonym: amidosulfonic acid,amidosulfuric acid,aminosulfonic acid,imidosulfonic acid,sulfamidic acid,sulfaminic acid,sulphamic acid,jumbo,aminosulfuric acid,sulphamidic acid ChEBI: CHEBI:9330 IUPAC Name: sulfamic acid SMILES: NS(=O)(=O)O
Sulfamic Acid, ACS Grade, 99.3 to 100.3%, LabChem™
CAS: 5329-14-6 Molecular Formula: H3NO3S Molecular Weight (g/mol): 97.088 InChI Key: IIACRCGMVDHOTQ-UHFFFAOYSA-N Synonym: amidosulfonic acid,amidosulfuric acid,aminosulfonic acid,imidosulfonic acid,sulfamidic acid,sulfaminic acid,sulphamic acid,jumbo,aminosulfuric acid,sulphamidic acid PubChem CID: 5987 ChEBI: CHEBI:9330 IUPAC Name: sulfamic acid SMILES: NS(=O)(=O)O
Sulfamic Acid, ACS Reagent Grade, Ricca Chemical
CAS: 5329-14-6 Molecular Formula: H3NO3S InChI Key: IIACRCGMVDHOTQ-UHFFFAOYSA-N PubChem CID: 5987 ChEBI: CHEBI:9330 IUPAC Name: sulfamic acid
Sulfamic Acid, ACS Reagent, Reagents
CAS: 5329-14-6 Molecular Formula: H3NO3S Molecular Weight (g/mol): 97.09 InChI Key: IIACRCGMVDHOTQ-UHFFFAOYSA-N Synonym: Amidosulfonic Acid, Aminosulfonic Acid, Sulphamic Acid IUPAC Name: sulfamic acid SMILES: NS(O)(=O)=O
Sulfamic Acid, Technical Contains 0.3% Anti-caking Agent, Reagents
Sulfamic Acid, Technical Contains 0.3% Anti-caking Agent, Technical, for general laboratory use. Manufactured in ISO 9001 facility.