missing translation for 'onlineSavingsMsg'
Learn More

Sulfamic Acid, Analytical Standard, 99.0-101.0% (T), MilliporeSigma™ Supelco™

Catalog No. 111019508
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019508 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111019508 Supplier Merck Emd Millipore Supplier No. 11358100MG

missing translation for 'validMainframeMsg'

Specifications

CAS 5329-14-6
Physical Form Neat
Linear Formula NH2SO3H
MDL Number MFCD00011603
Melting Point 215°C to 225°C (decomposition)
Synonym Amidosulfonic acid
Quantity 100 mg
Formula Weight 97.09
Percent Purity 99.0 to 101.0% (T)
Grade Analytical Standard