missing translation for 'onlineSavingsMsg'
Learn More

Acid Starch Indicator, Reagents

Catalog No. 01336013
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01336013 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01336013 Supplier Labchem Inc Supplier No. CA016100500A5
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Acid Starch Indicator, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Acid Starch Indicator
CAS 5329-14-6, 9005-84-9
Synonym Sulfamic Acid/Starch
Grade Reagent
Packaging Poly Bottle
Specific Gravity 2.0236
Color White to Off-white
Quantity 500 g
Physical Form Solid