missing translation for 'onlineSavingsMsg'
Learn More

Sulfamic Acid, Analytical Standard (for Acidimetry), ACS reagent, 99.3-100.3% (dried material), MilliporeSigma™ Supelco™

Catalog No. 111020395
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111020395 100 g
111019032 500 g
111018575 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 111020395 Supplier Merck Emd Millipore Supplier No. 86040100G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 5329-14-6
Linear Formula NH2SO3H
Technique Acidimetry
MDL Number MFCD00011603
Melting Point 215°C to 225°C (decomposition)
Synonym Amidosulfonic acid
Quantity 100 g
Formula Weight 97.09
Percent Purity 99.3 to 100.3% (dried material)
Grade Analytical Standard