missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfamic acid, 99%
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | White |
| Melting Point | 200.0°C to 208.0°C |
| Packaging | Plastic Bottle |
| Assay Percent Range | 99% |
| Linear Formula | H2NSO3H |
| Quantity | 250 g |
| Physical Form | Crystals or Crystalline Powder |
| Fieser | 11117 |
| Merck Index | 159053 |
| Solubility Information | Solubility in water: 146.8g/L (20°C). Other solubilities: sparingly soluble in alcohol and methanol,slightly soluble in acetone,insoluble in ether |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Warning |
| EINECSNumber | 226-218-8 |
RUO – Research Use Only