missing translation for 'onlineSavingsMsg'
Learn More

Sulfamic Acid, Technical Contains 0.3% Anti-caking Agent, Reagents

Catalog No. 01337569
Change view
Click to view available options
Quantity:
500 g
2.5 kg
10 kg
50 lb.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01337569 10 kg
01337568 2.5 kg
01337567 500 g
01337566 50 lb.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 01337569 Supplier Labchem Inc Supplier No. C211433310KG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Sulfamic Acid, Technical Contains 0.3% Anti-caking Agent, Technical, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Sulfamic Acid, Technical Contains 0.3% Anti-caking Agent
Color White to Off-White
CAS 5329-14-6
Packaging Poly Pail
Specific Gravity 2.15
Quantity 10 kg
Molecular Formula NH2SO3H
Physical Form Solid
Synonym Amidosulfonic Acid, Aminosulfonic Acid, Sulphamic Acid
Molecular Weight (g/mol) 97.0937
Grade Technical
Show More Show Less