Filtered Search Results
Ansell TouchNTuff™ 92-600 Disposable Chemical Resistant Nitrile Gloves
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Soft nitrile disposable gloves provide high levels of comfort and splash resistance against hazardous chemicals.
| Applications | Industrial |
|---|---|
| Product Type | Chemical-resistant Gloves |
| Color | Green |
| Latex Free | Yes |
| Material | Nitrile |
Ansell TouchNTuff™ 92-675 Industrial Disposable Nitrile Gloves
Disposable 5 mil thick nitrile gloves provide strength and durability for food processing and heavy-duty jobs.
| Tensile Strength | 14 MPa |
|---|---|
| Disposable | Single-use |
| Length (Metric) | 24 cm |
| Applications | Industrial,Laboratory |
| Finger Thickness (English) | 5.5 mil |
| Color | Blue |
| Length (English) | 9.5 in. |
| Material | Nitrile |
| Powdered | Powder Free |
| Finger Thickness (Metric) | 0.14 mm |
| Certifications/Compliance (Europe) | EN 374-1:2016, EN 374:2003, EN 420:2003 + A1:2009, EN 374-5:2016 |
| Thickness (English) | 5.5 mil |
| Palm Thickness (English) | 4.3 mil |
| Exterior Finish | Partially Textured |
| Certifications/Compliance | FDA 21 CFR 177-2600,ISO 9001 |
| AQL | 4 |
| Palm Thickness (Metric) | 0.11 mm |
Ansell TouchNTuff™ 69-318 Latex Gloves
Lightweight latex gloves feature textured fingers and tactile sensitivity for easy handling of small objects and delicate instruments.
| Tensile Strength | 18 MPa |
|---|---|
| Laboratory/Cleanroom Use | No |
| Disposable | Single-use |
| Length (Metric) | 24.1 cm |
| Applications | Laboratory,Industrial |
| Chemotherapeutic Rated | No |
| Finger Thickness (English) | 5.5 mil |
| Length (English) | 9.5 in. |
| Material | Latex |
| Powdered | Powder Free |
| Sterility | Non-sterile |
| Finger Thickness (Metric) | 0.14 mm |
| Thickness (Metric) | 0.14 mm |
| Accelerator Free | No |
| Series | TouchNTuff™ 69-318 |
| FDA 510 (k) | No |
| Product Type | Latex Disposable Gloves |
| Thickness (English) | 5.5 mil |
| Palm Thickness (English) | 4.7 mil |
| Latex Free | No |
| Exterior Finish | Partially Textured |
| AQL | 1.5 |
| Palm Thickness (Metric) | 0.12 mm |
Ansell AlphaTec™ 08-352 Neoprene Gloves, Knit Liner
Neoprene work gloves provide chemical resistance and offer a rough finish dip that makes handling wet, slippery materials easier and safer.
| Grip Design | Rough |
|---|---|
| Length (Metric) | 30.48 cm |
| EN388 Standard | EN 388:3121B, EN 388:3121 |
| Color | Green |
| Length (English) | 12 in. |
| Material | Neoprene |
| For Use With (Application) | Bulk chemical handling and transport, cleaning and de-greasing, handling alkalies, salts, oils, acids, caustics, alcohols and specialty chemicals |
| Thickness (Metric) | 0.38 mm |
| Series | AlphaTec™ 08-352 |
| EN ISO 374-1 | Type A |
| ANSI Abrasion Rating | ANSI 3 |
| Certifications/Compliance (Europe) | EN 420:2003 + A1:2009; Category III, EN 407 |
| EN ISO 374-1 CHEMICAL LETTER CODES | AKLMNPT |
| Thickness (English) | 15 mil |
| Latex Free | Yes |
| Lining | Lined |
| Liner Material | Interlock Cotton |
| Glove Cuff Style | Gauntlet |
Ansell KSR™ 22-515 Vinyl Coated Gloves with Interlock Knit Liner
These light, comfortable gloves are ideal for periods of extended wear during light-duty work. Vinyl coating enhances grip when working with small, difficult objects.
| Coating Material | PVC |
|---|---|
| EN388 Standard | 2121A |
| Color | Tan |
| Cut Resistance Rating | ANSI A1 |
| Material | Cotton |
| For Use With (Application) | Inspection, Assembly, and Metal Fabrication |
| Hand | Specific |
| Series | 22-515 |
| ANSI Abrasion Rating | ANSI 3 |
| Product Type | Dip-coated Gloves |
| Latex Free | Yes |
| Exterior Finish | No Texture |
| Liner Material | Cotton |
| Glove Cuff Style | Open |
Ansell AlphaTec™ 08-354 Neoprene Gloves, Interlocked Knit Lining
Liquid-proof industrial neoprene gloves offer a rough finish dip that makes handling wet, slippery materials easier and safer.
| Grip Design | Rough |
|---|---|
| Length (Metric) | 35.5 cm |
| EN 421 | No |
| EN388 Standard | EN 388:3121B, EN 388:3121 |
| Color | Green |
| Length (English) | 14 in. |
| Material | Neoprene |
| Thickness (Metric) | 0.76 mm |
| Series | AlphaTec™ 08-354 |
| EN ISO 374-1 | Type A |
| ANSI Abrasion Rating | ANSI 3 |
| Certifications/Compliance (Europe) | EN 420:2003 + A1:2009; Category III, EN 407:X1XXXX |
| EN ISO 374-1 CHEMICAL LETTER CODES | AKLMNPT |
| EN ISO 374 - 5 | Bacteria, Fungi only |
| Thickness (English) | 30 mil |
| Latex Free | Yes |
| Lining | Lined |
| Liner Material | Interlock Cotton |
| Glove Cuff Style | Gauntlet |
Ansell ActivArmr™ 27-600 Nitrile Coated Heavy Duty Work Gloves - Knitwrist
Gloves feature a unique coating formulation that provides increased resistance against sharp and abrasive environments, in addition to oil and grease.
| Construction | Cut and Sewn |
|---|---|
| Coating Material | Nitrile |
| EN388 Standard | 4121B |
| Color | Blue,Natural |
| Cut Resistance Rating | ANSI A1 |
| Material | Cotton |
| For Use With (Application) | Automotive Assembly, Machinery/Equipment Operation, and Metal Fabrication |
| Hand | Specific |
| Series | ActivArmr™ 27-600 |
| ANSI Abrasion Rating | ANSI 4 |
| Certifications/Compliance (Europe) | EN 420:2003 + A1:2009 |
| Product Type | Dip-coated Gloves |
| Latex Free | Yes |
| Glove Cuff Style | Knit |
Ansell ActivArmr™ 27-602 Fully Nitrile Coated Heavy Duty Cotton Jersey Gloves
Ansell's unique nitrile coating formulation offers resistance to sharp/abrasive materials, oil, and grease. The heavy duty design is durable, and certified clean for metals and motor manufacturing.
| Series | 27-602 |
|---|---|
| ANSI Abrasion Rating | ANSI 4 |
| Construction | Cut and Sewn |
| Product Type | Dip-coated Gloves |
| Coating Material | Nitrile |
| Color | Blue,Natural |
| Cut Resistance Rating | ANSI A1 |
| Latex Free | Yes |
| Material | Cotton |
| Exterior Finish | No Texture |
| For Use With (Application) | Automotive, Machinery/Equipment, and Metal Fabrication |
| Glove Cuff Style | Knit |
| Hand | Specific |
Ansell AlphaTec™ 29-865 Neoprene Chemical Resistant Gloves
Medium-duty unsupported neoprene gloves provide flexibility and ease of use.
| Chlorinated | Yes |
|---|---|
| Grip Design | Sandpatch |
| Length (Metric) | 33 cm |
| EN 421 | No |
| Applications | Industrial |
| EN388 Standard | EN 388:1000X, EN 388: 3111 |
| Color | Black |
| Length (English) | 13 in. |
| Material | Neoprene |
| For Use With (Application) | Automotive, Chemical, Janitorial, Sanitation |
| Thickness (Metric) | 0.43 mm |
| Series | AlphaTec™ 29-865 |
| EN ISO 374-1 | Type B |
| ANSI Abrasion Rating | ANSI 3 |
| Certifications/Compliance (Europe) | CE 0493, EN 420:2003 + A1:2009; Category III, EN 1149 |
| EN ISO 374-1 CHEMICAL LETTER CODES | AKL |
| EN ISO 374 - 5 | Bacteria, Fungi only |
| Thickness (English) | 17 mil |
| Model | 29-865 |
| Latex Free | Yes |
| Lining | Lined |
| Certifications/Compliance | 21 CFR 177-2600 |
| Liner Material | Cotton Flock |
| Glove Cuff Style | Straight |
Ansell Hynit™ 32-105 Light Duty Nitrile Coated Liquid-Repellent Gloves
Fully impregnated with an unique liquid repellent nitrile formulation to provide durability in abrasive environments, while also protecting workers against oil, grease, and dirt.
| Series | 32-105 |
|---|---|
| ANSI Abrasion Rating | ANSI 3 |
| Product Type | Dip-coated Gloves |
| Coating Material | Nitrile |
| Color | Blue |
| Cut Resistance Rating | ANSI A1 |
| Latex Free | Yes |
| Material | Cotton |
| Exterior Finish | No Texture |
| Liner Material | Cotton |
| For Use With (Application) | Automotive, Machinery/Equipment, Metal Fabrication, and Mining |
| Glove Cuff Style | Open |
| Hand | Specific |
Ansell AlphaTec™ Solvex™ 37-145 Nitrile Gloves
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Solvex™ 11 mil nitrile gloves are tactile and durable and can be used in a diverse range of applications including automotive, machinery/equipment and chemical.
| Chlorinated | Yes |
|---|---|
| Grip Design | Sandpatch |
| Length (Metric) | 33 cm |
| EN 421 | No |
| EN388 Standard | EN 388:4001, EN 388:3001X |
| Color | Green |
| Length (English) | 13 in. |
| Cut Resistance Rating | ANSI A4 |
| Material | Nitrile |
| For Use With (Application) | Chemical plants and transportation, chemical processing, Oil refining and Petrochemicals, cleaning and de-greasing, exposure to chemicals, Solvents, hazardous liquids, PVC cements or standing Water, janitorial and maintenance work |
| Thickness (Metric) | 0.27 mm |
| EN ISO 374-1 | Type A |
| ANSI Abrasion Rating | ANSI 5 |
| EN ISO 374-1 CHEMICAL LETTER CODES | JKNOPT |
| EN ISO 374 - 5 | Bacteria, Fungi only |
| Thickness (English) | 11 mil |
| Latex Free | Yes |
| Lining | Unlined |
| Certifications/Compliance | FDA 21 CFR 177.2600 |
| Glove Cuff Style | Gauntlet |
Ansell AlphaTec™ 13 in. Solvex™ 37-155 Nitrile Gloves
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Tactile and durable 15 mil nitrile gloves provide chemical resistance and comfort in wet or dry work environments across a diverse range of applications.
| Chlorinated | Yes |
|---|---|
| Grip Design | Sandpatch |
| Length (Metric) | 33 cm |
| EN 421 | No |
| EN388 Standard | EN 388:4101, EN 388:4101X |
| Color | Green |
| Length (English) | 13 in. |
| Material | Nitrile |
| For Use With (Application) | Chemical plants and transportation, chemical processing, Oil refining and Petrochemicals, cleaning and de-greasing, exposure to chemicals, Solvents, hazardous liquids, PVC cements or standing Water, janitorial and maintenance work |
| Thickness (Metric) | 0.38 mm |
| Series | AlphaTec™ Solvex™ 37-155 |
| EN ISO 374-1 | Type A |
| ANSI Abrasion Rating | ANSI 4 |
| Certifications/Compliance (Europe) | EN 420:2003 + A1:2009, Category III |
| EN ISO 374-1 CHEMICAL LETTER CODES | JKLOPT |
| EN ISO 374 - 5 | Bacteria, Fungi only |
| Thickness (English) | 15 mil |
| Latex Free | Yes |
| Lining | Unlined |
| Certifications/Compliance | FDA 21 CFR 177.2600 |
| Glove Cuff Style | Gauntlet |
Ansell AlphaTec™ 15 in. Solvex™ 37-165 Nitrile Gloves
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Heavy-duty 22 mil nitrile gloves offer versatile chemical protection that performs across a diverse range of applications.
| Chlorinated | Yes |
|---|---|
| Grip Design | Sandpatch |
| Length (Metric) | 38 cm |
| EN 421 | No |
| EN388 Standard | EN 388:4102, EN 388:4102X |
| Color | Green |
| Length (English) | 15 in. |
| Material | Nitrile |
| For Use With (Application) | Chemical plants and transportation, chemical processing, Oil refining and Petrochemicals, cleaning and de-greasing, exposure to chemicals, Solvents, hazardous liquids, PVC cements or standing Water, janitorial and maintenance work |
| Thickness (Metric) | 0.55 mm |
| Series | AlphaTec™ Solvex™ 37-165 |
| EN ISO 374-1 | Type A |
| ANSI Abrasion Rating | ANSI 5 |
| Certifications/Compliance (Europe) | CE 0493, EN 420:2003 + A1:2009, Category III |
| EN ISO 374-1 CHEMICAL LETTER CODES | AGJKLPT |
| EN ISO 374 - 5 | Bacteria, Fungi only |
| Thickness (English) | 22 mil |
| Latex Free | Yes |
| Lining | Unlined |
| Certifications/Compliance | FDA 21 CFR 177.2600 |
| Glove Cuff Style | Gauntlet |
Ansell AlphaTec™ SOLVEX™ 37-175 Nitrile Chemical Resistant Gloves
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Durable, comfortable chemical resistant gloves offer versatile hand protection.
| Chlorinated | Yes |
|---|---|
| Grip Design | Sandpatch |
| Length (Metric) | 33 cm |
| EN388 Standard | EN 388:4101X, EN 388:4101 |
| Finger Thickness (English) | 18.1 mil |
| Length (English) | 13 in. |
| For Use With (Application) | Automotive, Food Processing, Machinery, Chemicals, Utilities, Warehousing |
| Finger Thickness (Metric) | 0.45 mm |
| EN ISO 374-1 | Type A |
| EN ISO 374 - 5 | Bacteria, Fungi WITH Virus Protection |
| Thickness (English) | 17 mil |
| Palm Thickness (English) | 18.1 mil |
| Certifications/Compliance | FDA 21 CFR 177.2600,ASTM F1671 |
| Liner Material | Cotton Flock |
| Glove Cuff Style | Gauntlet |
| EN 421 | No |
| Applications | Industrial |
| Color | Green |
| Material | Nitrile |
| Thickness (Metric) | 0.46 mm |
| Series | AlphaTec™ Solvex™ 37-175 |
| ANSI Abrasion Rating | ANSI 4 |
| Certifications/Compliance (Europe) | CE 0086, EN 420:2003 + A1:2009, Category III |
| EN ISO 374-1 CHEMICAL LETTER CODES | JKLOPT |
| Model | 37-175 |
| Latex Free | Yes |
| Lining | Lined |
| Palm Thickness (Metric) | 0.45 mm |
Ansell AlphaTec™ 18 in. Solvex™ 37-185 Nitrile Gloves
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Heavy-duty, elbow length nitrile chemical resistant gloves suitable for a wide range of applications.
| Chlorinated | Yes |
|---|---|
| Grip Design | Sandpatch |
| Length (Metric) | 45.7 cm |
| EN 421 | No |
| Applications | Industrial |
| EN388 Standard | EN 388:4102X, EN 388:4102 |
| Color | Green |
| Length (English) | 18 in. |
| Material | Nitrile |
| For Use With (Application) | Chemical plants and transportation, chemical processing, Oil refining and Petrochemicals, cleaning and de-greasing, exposure to chemicals, Solvents, hazardous liquids, PVC cements or standing Water, janitorial and maintenance work |
| Thickness (Metric) | 0.55 mm |
| Series | AlphaTec™ Solvex™ 37-185 |
| EN ISO 374-1 | Type A |
| ANSI Abrasion Rating | ANSI 4 |
| Certifications/Compliance (Europe) | CE 0493, EN 420:2003 + A1:2009, Category III |
| EN ISO 374-1 CHEMICAL LETTER CODES | AGJKLPT |
| Product Type | Chemical-resistant Gloves |
| EN ISO 374 - 5 | Bacteria, Fungi only |
| Thickness (English) | 22 mil |
| Latex Free | Yes |
| Lining | Unlined |
| Certifications/Compliance | FDA 21 CFR 177.2600 |
| Manufacturing/QMS Audit Standards | ISO 18889 |
| Glove Cuff Style | Gauntlet |