missing translation for 'onlineSavingsMsg'
Learn More

Ansell AlphaTec™ 08-354 Neoprene Gloves, Interlocked Knit Lining

Catalog No. p-9012022
Change view
Click to view available options
Size:
10
9
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
11391120C 9
11391120D 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11391120C Supplier Ansell Supplier No. 0835411090
Only null left

Liquid-proof industrial neoprene gloves offer a rough finish dip that makes handling wet, slippery materials easier and safer.

  • Resists a wide range of chemicals, including oils, acids, solvents, and caustics
  • Ideal for applications involving petrochemicals, refining, cleaning, and degreasing electronics and materials handling
  • Flexible neoprene coating stretch provides comfortable fit
  • Two-piece lining eliminates seams, preventing hand irritation and premature wear-through
  • Textured finish provides secure grip on wet and slippery materials
  • Curved, preflexed fingers and wing thumb make working easier
  • Interlock knit lining allows easy donning and doffing
  • Gauntlet cuff

Specifications

ANSI Abrasion Rating ANSI 3
Certifications/Compliance (Europe) EN 420:2003 + A1:2009; Category III, EN 407:X1XXXX
Color Green
Latex Free Yes
Material Neoprene
Size 9
Thickness (English) 30 mil
Thickness (Metric) 0.76 mm
EN388 Standard EN 388:3121B, EN 388:3121
EN 421 No
EN ISO 374-1 Type A
EN ISO 374-1 CHEMICAL LETTER CODES AKLMNPT
EN ISO 374 - 5 Bacteria, Fungi only
For Use With (Application) Bulk chemical handling and transport, cleaning and de-greasing, handling Alkalies and Salts, handling Oils, Acids, Caustics and Alcohols, handling specialty chemicals
Glove Cuff Style Gauntlet
Grip Design Rough
Liner Material Interlock Cotton
Lining Lined
Model 8 354 9 212516
Series AlphaTec™ 08-354
Length (English) 14 in.
Length (Metric) 35.5 cm
Show More Show Less