missing translation for 'onlineSavingsMsg'
Learn More

Ansell AlphaTec™ 15 in. Solvex™ 37-165 Nitrile Gloves

Catalog No. p-9012019
Change view
Click to view available options
Size:
7
8
9
10
Catalog No. Size
11391138A 7
11391138B 8
11391138C 9
11391138D 10
4 options

Catalog No. 11391138A

Supplier: Ansell 3716511070

Only null left

Heavy-duty 22 mil nitrile gloves offer versatile chemical protection that performs across a diverse range of applications.

  • Nitrile construction protects against abrasions, cuts, snags and punctures
  • Curved finger design and contoured palm help reduce fatigue
  • Full cut for easy donning and doffing
  • Does not contain protein allergens
  • No inner cotton flocking, reducing risk of lint contamination
  • Sandpatch finish gives the glove a smooth surface
  • Fully reusable, with an unequaled abrasion resistance
  • Complies with FDA food handling requirements (21 CFR 177.2600)

Specifications

ANSI Abrasion Rating ANSI 5
Certifications/Compliance FDA 21 CFR 177.2600
Certifications/Compliance (Europe) CE 0493, EN 420:2003 + A1:2009, Category III
Chlorinated Yes
Color Green
Latex Free Yes
Material Nitrile
Size 7
Thickness (English) 22 mil
Thickness (Metric) 0.55 mm
EN388 Standard EN 388:4102, EN 388:4102X
EN 421 No
EN ISO 374-1 Type A
EN ISO 374-1 CHEMICAL LETTER CODES AGJKLPT
EN ISO 374 - 5 Bacteria, Fungi only
For Use With (Application) Chemical plants and transportation, chemical processing, Oil refining and Petrochemicals, cleaning and de-greasing, exposure to chemicals, Solvents, hazardous liquids, PVC cements or standing Water, janitorial and maintenance work
Glove Cuff Style Gauntlet
Grip Design Sandpatch
Lining Unlined
Model 37-165 7 117207
Series AlphaTec™ Solvex™ 37-165
Length (English) 15 in.
Length (Metric) 38 cm
Show More Show Less