missing translation for 'onlineSavingsMsg'
Learn More

Ansell KSR™ 22-515 Vinyl Coated Gloves with Interlock Knit Liner

Catalog No. p-8005313
Change view
Click to view available options
Size:
8
9
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
11391121A 8
11391121B 9
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11391121A Supplier Ansell Supplier No. 2251511080
Only null left

These light, comfortable gloves are ideal for periods of extended wear during light-duty work. Vinyl coating enhances grip when working with small, difficult objects.

  • Slip on cuff for easy donning and doffing
  • Ideal for long wear during light-duty applications
  • Enhanced grip for handling small, difficult objects

Specifications

ANSI Abrasion Rating ANSI 3
Coating Material PVC
Color Tan
Cut Resistance Rating ANSI A1
Exterior Finish No Texture
Hand Specific
Latex Free Yes
Material Cotton
Product Type Dip-coated Gloves
Size 8
EN388 Standard 2121A
For Use With (Application) Inspection, Assembly, and Metal Fabrication
Glove Cuff Style Open
Liner Material Cotton
Series 22-515
Show More Show Less