missing translation for 'onlineSavingsMsg'
Learn More

Ansell AlphaTec™ 13 in. Solvex™ 37-155 Nitrile Gloves

Tactile and durable 15 mil nitrile gloves provide chemical resistance and comfort in wet or dry work environments across a diverse range of applications.

$2.40 - $2.62

Specifications

ANSI Abrasion Rating ANSI 4
Certifications/Compliance FDA 21 CFR 177.2600
Certifications/Compliance (Europe) EN 420:2003 + A1:2009, Category III
Chlorinated Yes
Color Green
View More Specs

Products 5
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Quantity  
11-391-137A
View Documents
Ansell
3715511070
7
Pair for $2.40
Case of 144 Pair for $99.36
Only null left
 
11-391-137B
View Documents
Ansell
3715511080
8
Pair for $2.62
Case of 144 Pair for $100.80
Only null left
 
11-391-137C
View Documents
Ansell
3715511090
9
Pair for $2.62
Case of 144 Pair for $312.48
Only null left
 
11-391-137D
View Documents
Ansell
3715511100
10
Pair for $2.40
Case of 144 Pair for $100.80
Only null left
 
11-391-137E
View Documents
Ansell
3715511110
11
Pair for $2.40
Case of 144 Pair for $100.80
Only null left
 
Description

Description

Tactile and durable 15 mil nitrile gloves provide chemical resistance and comfort in wet or dry work environments across a diverse range of applications.

  • Nitrile construction protects against abrasions, cuts, snags and punctures
  • Curved finger design and contoured palm help reduce fatigue
  • Full cut for easy donning and doffing
  • Does not contain protein allergens
  • No inner cotton flocking, reducing risk of lint contamination
  • Sandpatch finish gives the glove a smooth surface
  • Fully reusable, with an unequaled abrasion resistance
  • Complies with FDA food handling requirements (21 CFR 177.2600)
Specifications

Specifications

ANSI 4
EN 420:2003 + A1:2009, Category III
Green
Nitrile
0.38 mm
No
JKLOPT
Chemical plants and transportation, chemical processing, Oil refining and Petrochemicals, cleaning and de-greasing, exposure to chemicals, Solvents, hazardous liquids, PVC cements or standing Water, janitorial and maintenance work
Sandpatch
AlphaTec™ Solvex™ 37-155
33 cm
FDA 21 CFR 177.2600
Yes
Yes
15 mil
EN 388:4101, EN 388:4101X
Type A
Bacteria, Fungi only
Gauntlet
Unlined
13 in.
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'