missing translation for 'onlineSavingsMsg'
Learn More

Ansell Hynit™ 32-105 Light Duty Nitrile Coated Liquid-Repellent Gloves

Catalog No. p-7219156
Change view
Click to view available options
Size:
8
9
10
Catalog No. Size
11391131A 8
11391131B 9
11391131C 10
3 options

Catalog No. 11391131A

Supplier: Ansell 3210511080

Only null left

Fully impregnated with an unique liquid repellent nitrile formulation to provide durability in abrasive environments, while also protecting workers against oil, grease, and dirt.

  • Special nitrile formulation combines flexibility with abrasion resistance
  • Liquid-repellent impregnation repels oil, grease, and dirt

Specifications

ANSI Abrasion Rating ANSI 3
Coating Material Nitrile
Color Blue
Cut Resistance Rating ANSI A1
Exterior Finish No Texture
Hand Specific
Latex Free Yes
Material Cotton
Product Type Dip-coated Gloves
Size 8
For Use With (Application) Automotive, Machinery/Equipment, Metal Fabrication, and Mining
Glove Cuff Style Open
Liner Material Cotton
Series 32-105
Show More Show Less