missing translation for 'onlineSavingsMsg'
Learn More

Ansell AlphaTec™ 18 in. Solvex™ 37-185 Nitrile Gloves
GREENER_CHOICE

Catalog No. p-9012021
Change view
Click to view available options
Size:
10
11
7
8
9
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
11391140A 7
11391140B 8
11391140C 9
11391140D 10
11391140E 11
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 11391140A Supplier Ansell Supplier No. 3718511070
Only null left

Heavy-duty, elbow length nitrile chemical resistant gloves suitable for a wide range of applications.

  • High performance nitrile compound
  • Combination of chemical resistance and strength ,for optimal results in wet or dry environments
  • Snag, puncture and abrasion protection compared with rubber or neoprene gloves
  • High levels of flexibility, comfort and dexterity
  • Suitable for heavy-duty cleaning applications
  • Complies with FDA food handling requirements (21 CFR 177.2600)

Specifications

ANSI Abrasion Rating ANSI 4
Certifications/Compliance FDA 21 CFR 177.2600
Certifications/Compliance (Europe) CE 0493, EN 420:2003 + A1:2009, Category III
Chlorinated Yes
Applications Industrial
Color Green
Latex Free Yes
Material Nitrile
Product Type Chemical-resistant Gloves
Size 7
Thickness (English) 22 mil
Thickness (Metric) 0.55 mm
EN388 Standard EN 388:4102X, EN 388:4102
EN 421 No
EN ISO 374-1 Type A
EN ISO 374-1 CHEMICAL LETTER CODES AGJKLPT
EN ISO 374 - 5 Bacteria, Fungi only
For Use With (Application) Chemical plants and transportation, chemical processing, Oil refining and Petrochemicals, cleaning and de-greasing, exposure to chemicals, Solvents, hazardous liquids, PVC cements or standing Water, janitorial and maintenance work
Glove Cuff Style Gauntlet
Grip Design Sandpatch
Lining Unlined
Manufacturing/QMS Audit Standards ISO 18889
Model 37-185 7 117298
Series AlphaTec™ Solvex™ 37-185
Length (English) 18 in.
Length (Metric) 45.7 cm
Show More Show Less