Learn More
Ahlstrom-Munksjo Standard Non Wet-Strengthened Qualitative Filter Paper, Grade 631
A medium speed grade used for the filtration of coarse particles and gelatinous precipitates such as ferric aluminum and magnesium hydroxides. It is also used in air monitoring and gas filtration when high flow rate is needed.
Supplier:  Ahlstrom-Munksjo 63100900
 Afficher plus de versions de ce produit
Numéro de catalogue. 09-790-67C
Il en reste null
Découvrez plus d'offres spéciales
Description
Description
Greener Choice Claims
  • Environmental benefits include:
    • Recyclable
Spécifications
Spécifications
10 μm
Filtration of coarse particles and gelatinous precipitates; air monitoring and gas filtration when high flow rate is needed
90 mm
Medium
631
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS