missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sorvall Legend 21 Every Lab Bundle PROMO

Bring your molecular work together by using the Every Lab Package.

Supplier:  Thermo Scientific™ 75002439PR

Numéro de catalogue. 01185012PM


Il en reste null
Découvrez plus d'offres spéciales

Description

Description

Covers basic and complex workflows

  • Our ventilated and refrigerated microcentrifuges reduce spin times by up to 20%, making them ideal for high-end molecular or genomic research applications
  • The Thermo Scientific™ included 24 X 1.5/2.0 mL rotor with ClickSeal™ biocontainment lid allows you to run tubes from 1.5 to 2.0 mL tube to mini-preps and spin columns
  • Pipettes cover a range of volumes to match necessary volumes, from 0.2-2 μL to 100-1000 μL
  • Snap Cap tubes offer versatility in the lab: Can be frozen or heated in a water bath

Save time with faster sample processing

  • Lightweight rotors provide acceleration up to 21,100 x g in 12 seconds.
  • Refrigerated models offer cooling from room temperature to set temperature in just 9 minutes.
  • Color coding of pipettes helps to select the correct pipette volume while working

Ideal for everyday laboratory use

  • High performance engineered polymer rotors are designed for outstanding chemical resistance to minimize corrosion from aggressive reagents
  • Conforms to the latest clinical and safety standards including 98/79/EC In Vitro Diagnostic Directive
  • Pipettes are fully autoclavable, minimizing disruption and downtime
  • Finnpipette pipettes are made of polyvinylidene fluoride (PVDF) components that stand up to harsh chemical, UV light and physical damage
Spécifications

Spécifications

24 x 1.5/2 mL
21,100 x g
Everylab Centrifuge Ventilated Package - Sorvall Legend 120 V, 21 V (including 24 x 1.5/2 mL rotor), F2 Single Channel Pipettes - GLP Kit 2, Snap Cap tubes (1 case 5000 tubes)
Sorvall
14,800 rpm
Microcentrifuge
No
120 V
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
FDS
Documents

Documents

Promotions

Promotions