Learn More
MilliporeSigma™ SeQuant™ ZIC™-HILIC Analytical PEEK Columns
PEEK coated HPLC column
Supplier:  MilliporeSigma™ 1504490001
 Afficher plus de versions de ce produit
Numéro de catalogue. M1504490001
Il en reste null
Découvrez plus d'offres spéciales
Description
Description
SeQuant™ ZIC™-HILIC is the ideal choice for separation of polar and hydrophilic compounds. The high-performance, truly zwitterionic stationary phase, ensures reproductible retention of all types of compounds that have poor retention and are difficult to separate on reversed-phase HPLC columns. ZIC™-HILIC is designed for compounds such as acids and bases, anions and cations, carbohydrates, metabolites, metal complexes, amino acids, peptides, protein digests and much more
Spécifications
Spécifications
4.6 mm
3.5 μm
200 Å
Metal-free PEEK
150 mm
200 Å
1/Pk.
Column
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS