missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Sephacryl™ High Resolution Media
Purify proteins, polysaccharides, and other macromolecules by gel filtration at laboratory and industrial scale. Cytiva Life Sciences™Sephacryl™ High Resolution Media enables fast and reproducible purifications.
$356.11 - $1479.99
Spécifications
| Buffer | 0.2M NaOH, 0.1M HCl, 1M Acetic Acid, 8M Urea, 6M Guanidine HCl, 1% SDS, 20% Ethanol, 30% Propanol, 30% Aceto nitrile (tested at 40°C for 1 week), 0.5M NaOH (only for cleaning-in-place) |
|---|---|
| Height (English) | 5.9 in. |
| Height (Metric) | 15cm |
| Max. Pressure | 1 bar |
| pH Range | 3 to 11 |
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Product Type | Flow Rate | Particle Size | Prix | Quantité | ||
|---|---|---|---|---|---|---|---|---|---|
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Product Type | Flow Rate | Particle Size | Prix | Quantité | ||
|
45-002-056
|
Cytiva
17060901 |
750 mL | Sephacryl S-400 HR media | >150 cm/hr. | 50μm |
chaque for $1,479.99
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
|
|
45-002-057
|
Cytiva
17059910 |
150 mL | Sephacryl S-300 HR media | >150 cm/hr. | 50μm |
chaque for $356.15
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
|
|
45-002-059
|
Cytiva
17058410 |
150 mL | Sephacryl S-200 HR media | >150 cm/hr. | 50μm |
chaque for $356.11
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
|
Description
- Fast and reproducible purification of proteins, polysaccharides, and other macromolecules
- Gel filtration media providing high resolution with fast and easy operation
- Available in five different fractionation ranges, allows for reproducible separations over a wide molecular weight range
- BioProcess medium supported for industrial applications and well-established in approved processes
- Available in different bulk pack sizes and convenient pre-packed formats for easy scale-up and process development
- Long-term physical, chemical stability and easy handling
- Hydrophilic nature enables maximum recovery and minimum nonspecific adsorption
- Average particle Size: 50μm
- Matrix: Allyl dextran and N, N'-methylenebisacrylamide
- pH stability working range: 3-11
- Shelf Life: 5years
Spécifications
| 0.2M NaOH, 0.1M HCl, 1M Acetic Acid, 8M Urea, 6M Guanidine HCl, 1% SDS, 20% Ethanol, 30% Propanol, 30% Aceto nitrile (tested at 40°C for 1 week), 0.5M NaOH (only for cleaning-in-place) | |
| 15cm | |
| 3 to 11 | |
| 4° to 30°C | |
| Gel Filtration Media |
| 5.9 in. | |
| 1 bar | |
| Allyl Dextran and N, N'-methylenebisacrylamide | |
| Purification of Proteins, Polysaccharides and other macromolecules |