missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Sephacryl™ High Resolution Media

Purify proteins, polysaccharides, and other macromolecules by gel filtration at laboratory and industrial scale. Cytiva Life Sciences™Sephacryl™ High Resolution Media enables fast and reproducible purifications.

Supplier:  Cytiva 17060901

 Afficher plus de versions de ce produit

Numéro de catalogue. 45-002-056

Il en reste null
Découvrez plus d'offres spéciales

Description

Description

  • Fast and reproducible purification of proteins, polysaccharides, and other macromolecules
  • Gel filtration media providing high resolution with fast and easy operation
  • Available in five different fractionation ranges, allows for reproducible separations over a wide molecular weight range
  • BioProcess medium supported for industrial applications and well-established in approved processes
  • Available in different bulk pack sizes and convenient pre-packed formats for easy scale-up and process development
  • Long-term physical, chemical stability and easy handling
  • Hydrophilic nature enables maximum recovery and minimum     nonspecific adsorption

  • Average particle Size: 50μm
  • Matrix: Allyl dextran and N, N'-methylenebisacrylamide
  • pH stability working range: 3-11
  • Shelf Life: 5years

Spécifications

Spécifications

0.2M NaOH, 0.1M HCl, 1M Acetic Acid, 8M Urea, 6M Guanidine HCl, 1% SDS, 20% Ethanol, 30% Propanol, 30% Aceto nitrile (tested at 40°C for 1 week), 0.5M NaOH (only for cleaning-in-place)
750 mL
15cm
1 bar
50μm
4° to 30°C
Gel Filtration Media
Sephacryl S-400 HR media
5.9 in.
>150 cm/hr.
3 to 11
Allyl Dextran and N, N'-methylenebisacrylamide
Purification of Proteins, Polysaccharides and other macromolecules
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
FDS
Documents

Documents

Promotions

Promotions