Learn More
Sebacoyl chloride, 95%
CAS: 111-19-3 | C10H16Cl2O2 | 239.136 g/mol
$137.27 - $1471.10
Identifiants chimiques
CAS | 111-19-3 |
---|---|
Molecular Formula | C10H16Cl2O2 |
Molecular Weight (g/mol) | 239.136 |
MDL Number | MFCD00000770 |
InChI Key | WMPOZLHMGVKUEJ-UHFFFAOYSA-N |
Synonym | sebacoyl chloride, sebacyl chloride, sebacoyl dichloride, sebacic acid dichloride, decanedioyl chloride, sebacoylchloride, octane-1,8-dicarbonyl chloride, sebacoyl chloride, cp, acmc-1byrk, decanedioic acid chlorideAfficher plus de résultats |
PubChem CID | 66072 |
IUPAC Name | decanedioyl dichloride |
SMILES | C(CCCCC(=O)Cl)CCCC(=O)Cl |
Numéro de catalogue | Numéro du manufacturier. | Quantity | Prix | Quantité | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Numéro de catalogue | Numéro du manufacturier. | Quantity | Prix | Quantité | ||||||
AAA1422614
|
Thermo Scientific Chemicals
A1422614 |
25 g |
chaque for $137.27
|
|
Vous avez déjà un compte ? Connectez-vous pour voir vos prix et disponibilités. Nouveau ici ? Inscrivez-vous dès aujourd'hui ! |
|||||
AAA1422622
|
Thermo Scientific Chemicals
A1422622 |
100 g |
chaque for $408.04
|
|
Vous avez déjà un compte ? Connectez-vous pour voir vos prix et disponibilités. Nouveau ici ? Inscrivez-vous dès aujourd'hui ! |
|||||
AAA1422636
|
Thermo Scientific Chemicals
A1422636 |
500 g |
chaque for $1,471.10
|
|
Vous avez déjà un compte ? Connectez-vous pour voir vos prix et disponibilités. Nouveau ici ? Inscrivez-vous dès aujourd'hui ! |
|||||
Description
Sebacoyl chloride is used in the synthesis of nylon-6,10 polymer by reacting with hexamethylenediamine. It is also used to prepare decanedioic acid bis-(3-dimethylamino-propyl) ester in the presence of triethylamine as catalyst. It is used as an intermediate in chemical synthesis. It is involved in the production of polyamide microcapsules by polycondensation of p-phenylenediamine with sebacoyl chloride using diethylenetriamine (DETA) cross-linker and oleic acid as a plasticizer.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsSebacoyl chloride is used in the synthesis of nylon-6,10 polymer by reacting with hexamethylenediamine. It is also used to prepare decanedioic acid bis-(3-dimethylamino-propyl) ester in the presence of triethylamine as catalyst. It is used as an intermediate in chemical synthesis. It is involved in the production of polyamide microcapsules by polycondensation of p-phenylenediamine with sebacoyl chloride using diethylenetriamine (DETA) cross-linker and oleic acid as a plasticizer.
Solubility
Miscible with hydrocarbons and ethers.
Notes
Store in a cool place. Moisture sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with water, acids, bases, amines, alcohols and oxidizing agents.
Identifiants chimiques
111-19-3 | |
239.136 | |
WMPOZLHMGVKUEJ-UHFFFAOYSA-N | |
66072 | |
C(CCCCC(=O)Cl)CCCC(=O)Cl |
C10H16Cl2O2 | |
MFCD00000770 | |
sebacoyl chloride, sebacyl chloride, sebacoyl dichloride, sebacic acid dichloride, decanedioyl chloride, sebacoylchloride, octane-1,8-dicarbonyl chloride, sebacoyl chloride, cp, acmc-1byrk, decanedioic acid chlorideAfficher plus de résultats | |
decanedioyl dichloride |
Spécifications
111-19-3 | |
1.121 | |
165°C (329°F) | |
C10H16Cl2O2 | |
MFCD00000770 | |
UN2922 | |
Moisture sensitive | |
Miscible with hydrocarbons and ethers. | |
C(CCCCC(=O)Cl)CCCC(=O)Cl | |
239.136 | |
239.14 | |
Sebacoyl chloride |
-5°C | |
161°C (14 mmHg) | |
Acrid | |
1.468 | |
25 g | |
1365665 | |
sebacoyl chloride, sebacyl chloride, sebacoyl dichloride, sebacic acid dichloride, decanedioyl chloride, sebacoylchloride, octane-1,8-dicarbonyl chloride, sebacoyl chloride, cp, acmc-1byrk, decanedioic acid chloride | |
WMPOZLHMGVKUEJ-UHFFFAOYSA-N | |
decanedioyl dichloride | |
66072 | |
97% |
Sécurité et manipulation
GHS H Statement
H310-H314-H318-H302
Fatal in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
P260-P262-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H302-H310-H314-H335
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.
missing translation for 'einecsNumber' : 203-843-4
missing translation for 'rtecsNumber' : HD8454250
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only